Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group A Antigen Monoclonal Antibody (HE-193)
GREENER_CHOICE

Catalog No. 01676718 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-718 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-718 Supplier Invitrogen™ Supplier No. MA119693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes human blood group A (monofucosyl and difucosyl A antigens with chain types 1 and 2, A antigens with chain types 3, 4, 5, 6) and Forssman antigen.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group A Antigen
Applications Agglutination, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HE-193
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with no preservative
Gene ABO
Gene Accession No. P16442
Gene Alias A transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; B transferase; B(A) alpha-1,3-galactosyltransferase; Blood Group A1; Blood Group A2; Blood Group A3; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group B transferase; NAGAT; Transferase; transferase A
Gene Symbols ABO
Host Species Mouse
Immunogen Mixture of erythrocytes from saliva of secretors with blood group A.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.