Learn More
Description
Human blood group A antigen belongs to a group of carbohydrate determinants carried on both glycolipids and glycoproteins; it is detected on erythrocytes and certain epithelial cells. The antibody HE-195 recognizes human blood group A including weak variants A3 , AX , A3B, AXB. The specifity of antibody HE-195 was confirmed by comparison of specifity and reactivity to standard reagent using >5.000 samples of blood. Agglutination test A1, A1B, A2B.
Specifications
Specifications
| Antigen | Blood Group A Antigen |
| Applications | Agglutination |
| Classification | Monoclonal |
| Clone | HE-195 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with <0.1% sodium azide |
| Gene | ABO |
| Gene Accession No. | P16442 |
| Gene Alias | A transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; B transferase; B(A) alpha-1,3-galactosyltransferase; Blood Group A1; Blood Group A2; Blood Group A3; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group B transferase; NAGAT; Transferase; transferase A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
