Learn More
Invitrogen™ Blood Group A1, A2 Antigen Monoclonal Antibody (87-G)

Description
MA1-7671 detects A1, A2 human blood protein from human samples. MA1-7671 has been successfully used in ELISA, agglutination and inhibition assays. The MA1-7671 immunogen is A1, A2 human blood protein.
Specifications
Specifications
| Antigen | Blood Group A1, A2 Antigen |
| Applications | Agglutination, ELISA, Inhibition Assays |
| Classification | Monoclonal |
| Clone | 87-G |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Antibody with 0.3M NaCl, 1.25mM EDTA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | Blood Group A |
| Host Species | Mouse |
| Immunogen | A1, A2 human blood protein. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.