Learn More
Description
This antibody reacts with human blood group B. The specifity of the antibody HEB-29 was confirmed by comparison of specifity and reactivity to standard reagent using >5000 samples of blood.
Specifications
Specifications
| Antigen | Blood Group B Antigen |
| Applications | Agglutination, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HEB-29 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with no preservative |
| Gene | ABO |
| Gene Accession No. | P16442 |
| Gene Symbols | ABO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
