Learn More
Invitrogen™ Blood Group B Antigen Monoclonal Antibody (Z5H-2)

Description
MA1-7666 detects B human blood protein from human samples. MA1-7666 has been successfully used in agglutination applications. The MA1-7666 immunogen is B human blood protein. Store short-term at 4ºC, long-term at -80ºC.
Specifications
Specifications
| Antigen | Blood Group B Antigen |
| Applications | Agglutination |
| Classification | Monoclonal |
| Clone | Z5H-2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Antibody with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2 |
| Gene | ABO |
| Gene Accession No. | P16442 |
| Gene Symbols | ABO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.