Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Blood Group M Antigen Monoclonal Antibody (GH-9)
GREENER_CHOICE

Catalog No. 01670410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-410 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-410 Supplier Invitrogen™ Supplier No. MA17676
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7676 detects M human blood protein from human samples. MA1-7676 has been successfully used in agglutination applications. The MA1-7676 immunogen is a M human blood protein.

The MNS antigen system is a human blood group system based upon two genes (glycophorin A and glycophorin B) on chromosome 4. The MN blood group system is under the control of an autosomal locus found on chromosome 4, with two alleles designated L-M and L-N. The blood type is due to a glycoprotein present on the surface of red blood cells, which behaves as a native antigen. Phenotypic expression at this locus is codominant because an individual may exhibit either one or both antigenic substances. These antigens were an early discovery and are some of the oldest blood antigens known after the ABO system. Anti-M and anti-N antibodies are usually IgM and are rarely associated with transfusion reactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group M Antigen
Applications Agglutination
Classification Monoclonal
Clone GH-9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin, 0.5% saccharose and 0.01% sodium azide; pH 7.2
Gene Alias Blood Group LM; MN Blood Group
Host Species Mouse
Immunogen M human blood protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.