Learn More
Biopac Systems Inc BLOOD PRESSURE TRANSDUCER
SDP
Supplier:  Biopac Systems Inc TSD104A
Encompass
measures direct arterial or venous blood pressure in animals or records pressure changes within an in-vitro circuit e.g. Langendorff heart preparation
Catalog No. NC2164648
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.