Learn More
Description
Use hands-on activities to understand the nature and importance of antigen-antibody reactions.
- Provide accurate simulation of the blood typing procedure
- No toothpicks, stirring or waiting for results
- Kit contains materials for 10 groups
Kits use simulated blood only.
Specifications
Specifications
| Certifications/Compliance | 49 CFR 173.4 |
| Color | White |
| Product Type | Blood Type Kit |
| Grade | 7 to 14 |
| Age | 13+ years |
| For Use With (Application) | Simulated abo/rh blood typing |
| Includes | 4 simulated blood samples, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1bottle simulated anti-rh serum, 40 blood typing trays, Teacher's manual and student study guide copymasters |
| Quantity | 1 Ea. |
| Class Size | 10 Lab Groups |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
