Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

First Aid Only™ Bloodborne Pathogen Spill Cleanup Kit
SDP

Catalog No. 18366636 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
10 x 2 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
18-366-636 10 x 2 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-636 Supplier First Aid Only™ Supplier No. PLS2045
Add to Cart
Add to Cart

Safely clean up messy spills of blood or bodily fluids using this OSHA-compliant kit packed with 18 essential supplies.

  • OSHA-compliant Bloodborne Pathogen kit includes 18 essential supplies for cleaning up a bloodborne-pathogen or bodily-fluid spill
  • Personal Protection Equipment helps minimize exposure of personnel to potential contaminants
  • Safely clean up and dispose of potentially infectious bodily fluids such as vomit, blood, urine or feces
  • Disposable kit is single-use to prevent cross contamination
  • Includes: 2 - pairs of nitrile exam gloves, 1 - 8 x 8 in. germicidal wipe, 1 - scoop/spatula, 1 - 2 oz. pouch of spill clean-up powder, 3 - disposable clean-up towels, 2 - clear bags, 1 - twist tie, 1 - bodily fluid pick-up guide, 3 - antiseptic towelettes, 1 - eye and face shield, 1 - disposable gown, 1 - 24 x 24 in., 10 gal. capacity biohazard bag

Specifications

Certifications/Compliance OSHA
Material Polyethylene
Type Spill Kit
Dimensions (L x W x H) 10 x 2 x 12 in.
Weight (English) 0.5 lb.
Weight (Metric) 227 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.