Description
- Convenient Tear-open box for quick access
- Includes 1 control gown
- Includes 1 eye and face shield or mask
- Includes 2 exam gloves
- Includes 1 biohazard bag
- Includes 2 BZK antiseptic wipes
- Designed for spill cleanup applications
- Suitable for standalone First-aid use or as a kit refill
- Includes essential barrier protection for safer handling
- Gloves help minimize direct contact with potentially infectious materials
- Mask and protective eyewear help protect the face and eyes from exposure
- Disposable coverings provide an additional protective barrier
- Helps reduce exposure to blood and other potentially infectious materials
Specifications
Specifications
| Size | Universal |
| Color | White, Blue |
| Product Type | Safety Pack |
| Sterility | Non-sterile |
| Disposable | Single-use |