Learn More
Medique BloodStopper Compress Wrap
Catalog No. 19090584 Shop All Medique Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-090-584 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-584 Supplier Medique Supplier No. 64101
Add to Cart
Add to Cart
This item is not returnable. View return policy
Conforming sterile wrap to control severe bleeding.
  • Super-absorbent non-stick pad
  • Removable gauze rolls for compression
  • Can be used as sling to secure splint
Specifications
Absorbancy Super Absorbent Non-stick Pad
Product Type Blood Stopper Compress Wrap
For Use With (Application) Control severe bleeding
Sterility Sterile
Includes Removable gauze rolls for compression
Quantity 1 Ea.
Dimensions (L x W) 9 x 5 in.