Learn More
Bullard™ Blower Assembly for (EVAHL) PAPR System
SDP
Designed to be used with the EVAHL Powered Air Purifying Respirator
Supplier:  Bullard™ EVAHL4DB
 View more versions of this product
Catalog No. 17-989-540
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
Description
Description
  • Choice of assembly configurations
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.
Specifications
Specifications
4 to 12 hr.
Lithium Polymer
NIOSH, ANSI/ISA 2.12.01-2007, CSA approvals for Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
Blower, vinyl decon belt, battery and HE filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.