Learn More
Description
- Plastic tags resist the elements while providing a long lasting identification solution Multiple color options are available for color-coding purposes
- Blank plastic lock-on tags have tab that loops into itself, effectively allowing you to fasten the tag without any additional materials
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor or outdoor use
Specifications
Specifications
| Quantity | 25/Pk. |
| Color | Blue |
| Material | Plastic |
| Includes | (25) nylon fasteners |
| Shape | Rectangle |
| Dimensions (L x W) | 2.5 x 0.5 in. |
| Label Type | Not laminated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.