Learn More
Educational Innovations Fluorescent Blue Magic Sand 1lb
SDP
Supplier:  Educational Innovations MS2BLU
Encompass
Magic Sand is regular sand that has been dyed and coated with a hydrophobic material - a substance that repels water. The coating on the outside of the magic sand repels water and keeps the sand dry! Simply remove the sand from the water with a spoon... it is perfectly dry! Available in three fluorescent colors. Comes in one pound packages.
Catalog No. NC0458372
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.