Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Blue Light Converter Screen
GREENER_CHOICESDP

Catalog No. 01257179 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Gbox Mini, Nugenius, Ingenius
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-257-179 Gbox Mini, Nugenius, Ingenius
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-179 Supplier Syngene™ Supplier No. CONVERTBLUE2126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blue Light Converter Screen converts UV to blue light for imaging of Safe Dyes. Use with UV transilluminators.

  • For use with Gbox Mini, NuGenius, Ingenius.

Specifications

Product Line Syngene
For Use With (Application) UV to blue light conversion
Product Type Convertor Screen
Quantity 1 Ea.
Color Blue
For Use With (Equipment) Gbox Mini, Nugenius, Ingenius
Size 21 x 26 cm
Voltage 100/115/220/230 V
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.