Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Blue Litmus pH Test Paper: Acid Indicator
SCI_ED

Catalog No. S80845 Shop All Eisco Products
Change view
Click to view available options
pH Range:
4.3 to 6.8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S80845 4.3 to 6.8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80845 Supplier Eisco™ Supplier No. LITMUSBLUEPK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicates the presence of an acid with a pH between 4.3-6.8

  • Blue Litmus Paper pH Tests
  • Indicates the presence of acid
  • pH range from 4.3-6.8
  • 100 Strips/Vial, 12 Vials/Pk

Specifications

pH Range 4.3 to 6.8
For Use With (Application) Detecting Acidity
Type Litmus Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.