Description
The BD FACSMelody™ BRYG 8 Color Plate Cell Sorter with newly added 4-way cell sorting:
- Makes the complex world of flow cytometry and sorting accessible to more researchers
- Combines the proven and exclusive BD technology of high-end sorters with automation and simplified software
- Helps in obtaining deep scientific insights, lab efficiency and cost savings
- Automated instrument setup eliminates or simplifies routine tasks
- High sensitivity and resolution to detect and sort dim cells and rare cells and resolve adjacent populations such as low-antigen density cells, small particles and DNA cell cycles
- BD CS and T technology ensures consistent instrument performance and reliability
- Sort performance:
- Droplet sorting
- 34,000 drops per second
- Automated setup, optimization and monitoring of dropletbreakoff and sort streams
- Automated drop-delay determination with BD FACS™ Accudrop beads
- Automated clog detection and sort tube protection system using Sweet Spot technology
Specifications
Specifications
| Dimensions (D x W x H) | Cell Sorter: 49.5 x 55.9 x 48.3 cm (19.5 x 22 x 19 in.); Electronics Box: 50.8 x 55.9 x 48.3 cm (20 x 22 x 19 in.) |
| Voltage | 100/115/230 V |
| Product Type | Flow Cytometry Cell Sorter |
| Laser Features | 3 Lasers (488 nm blue direct diode laser, output power: 20 mW, nominal power: 16 mW; 640 nm red direct diode laser, output power: 40 mW, nominal power: 36 mW; 561 nm yellow-green optically pumped semiconductor laser, output power 50 mW, nominal power: 40 mW) |
| Includes | Starter Kit |