Learn More
Leica Biosystems BLUE RIBBON PARAFFIN 10X2LB/CS
Supplier:  Leica Biosystems 3801360
Encompass_Preferred
Blue Ribbon is a universal paraffin containing synthetic polymers that can be used both as an infiltration and embedding medium. The paraffin enables the impregnation of tissue to occur rapidly and completely. The wax however will remain stable even at elevated temperatures. It is available in a case of 10 x 2 lb (0.91 kg) lightweight bags to facilitate handling. The bluish tint promotes better visibility of smaller or milky-colored/opaque specimens popular with Dermatology and Urology Labs.
Catalog No. NC1180943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.