Learn More
Lascar Electronics Inc Temperature & Humidity Bluetooth Data Logger
SDP
Supplier:  Lascar Electronics Inc ELBT2
Encompass_Preferred
The EL-BT-2 data logger measures and stores up to 500,000 temperature and humidity readings. The LCD screen on the device indicates Bluetooth connection status as well as logger operation.
Catalog No. NC1856632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.