Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLVRB/Biliverdin reductase B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570788
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-157-0788 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0788 Supplier Bethyl Laboratories, Inc Supplier No. A305570AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).

Specifications

Antigen BLVRB/Biliverdin reductase B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene BLVRB
Gene Accession No. P30043
Gene Alias BVRB, FLR, SDR43U1, BVR-B, FR, GHBP, NADPH-dependent diaphorase, NADPH-flavin reductase, biliverdin-IX beta-reductase, flavin reductase (NADPH), green heme-binding protein, short chain dehydrogenase/reductase family 43U, member 1
Gene Symbols BLVRB
Host Species Rabbit
Immunogen Between 156 and 206
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.