Learn More
Charles River Labs Cell Solutions CD34+ Stem/Progenitor Cells, 500,000 cells, Cryopreserved
Supplier:  Charles River Labs Cell Solutions BM34C3
Encompass
CD34+ stem cells are multipotent and can give rise to all cell types in blood. While compromising only 1.5% of bone marrow mononuclear cell population, CD34+ stem cells are responsible for all lymphohematopoietic lineages and are now commonly used clinically as they have been shown to provide long-term durable donor-derived host lymphohematopoietic reconstitution. CD34+ cells are isolated using positive immunomagnetic cell separation procedures from bone marrow.
Catalog No. 50-225-6123
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.