Learn More
Abcam BML-210 HDAC Inhibitor, Solid, Purity >98%, 5 mg
Description
- Molecular Weight: 339.4 Da
- Purity: >98%
- Form: Solid
- Activity: Histone deacetylase (HDAC) inhibitor (IC50 = 87 μM)
- Key Functions: Induces changes in gene expression profile; demonstrates antiproliferative activity in vitro
- Storage Buffer: 100% N'-(2-aminophenyl)-N-phenyloctanediamide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Histone Deacetylase |
| Buffer | 100% N'-(2-aminophenyl)-N-phenyloctanediamide |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% N'-(2-aminophenyl)-N-phenyloctanediamide |
| Quantity | 5 mg |
| For Use With (Application) | Histone deacetylase inhibition, Gene expression regulation, Antiproliferative studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.