Learn More
Enzo Life Sciences BML-277, (10mg), CAS Number: 516480-79-8
SDP
Supplier:  Enzo Life Sciences BMLEI3880010
Encompass_Preferred
Chk inhibitor. BML-277 is a highly selective inhibitor of Chk2 (IC50=15nM). It displays potent radioprotective activity and prevents apoptosis of human T cells subjected to ionizing radiation (EC50= 3-7.6µM). Useful tool for dissecting the role of Chk2 in cellular signaling. Alternative Name: Chk2 Inhibitor II. Formula: C20H14ClN3O2. MW: 363.8. CAS Number: 516480-79-8. Purity: ≥98% (HPLC). Appearance: Off-white solid. Solubility: Soluble in DMSO (20mg/ml) or ethanol (20mg/ml). Long Term Storage: -20°C
Catalog No. NC1076106
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.