Learn More
Apexbio Technology LLC BMN-673 8R,9S 1207456-00-5 10mg
SDP
Supplier:  Apexbio Technology LLC A324610
Encompass_Preferred
BMN-673 8R 9S (CAS 1207456-00-5) is an inactive stereoisomer of BMN-673 a potent poly(ADP-ribose) polymerase 1 and 2 (PARP1/2) inhibitor While BMN-673 demonstrates selective cytotoxicity toward tumor cells with deficiencies in BRCA1 BRCA2 or PTEN exploiting vulnerabilities in DNA repair pathways BMN-673 8R 9S lacks inhibitory activity This renders BMN-673 8R 9S a valuable negative control in studies investigating PARP inhibition mechanisms and differentiating on-target from off-target effects in cellular and in vivo models of DNA repair deficiency Its use supports rigorous evaluation of PARP1/2 inhibitor specificity in translational cancer research
Catalog No. 50-000-95805
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.