Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BMP-2 Quantikine ELISA Kit
GSA_VASDP

Catalog No. DBP200 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DBP200 1 Plate
SBP200 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DBP200 Supplier R&D Systems™ Supplier No. DBP200

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BMP-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The BMP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat BMP-2 in cell culture supernates (50 ul), bone extracts (50 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 29pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Bone Extracts, Cell Culture Supernatant
Specificity Natural and mammalian-expressed recombinant human BMP-2. This assay does not recognize E. coli-expressed BMP-2.
Sufficient For 1 x 96-well Plate
Target Species Human, Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 650 (Human); 12156 (Mouse); 29373 (Rat)
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Bone Extracts (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BDA2, BMP2, BMP-2, BMP-2 A, BMP2ABone morphogenetic protein 2 A, bone morphogenetic protein 2, SSFSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.