Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537844
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537844 Supplier Invitrogen™ Supplier No. PA537844
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BMP1
Gene Accession No. P13497, P98063
Gene Alias BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD
Gene Symbols BMP1
Host Species Goat
Immunogen Peptide with sequence C-DTIVPKYEVNGVK, from the internal region of BMP1 (aa284-296).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12153, 649, 83470
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.