Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BMP51/BMP53 Label Maker Cartridge, Reflective Tape (B-584)

Catalog No. 19107901 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
Yellow
Width (English):
1 in.
1.5 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (English)
19-107-901 White 1.5 in.
19-107-899 White 1 in.
19-107-900 Yellow 1 in.
19-107-902 Yellow 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-107-901 Supplier Brady™ Supplier No. MC1500584

To print a variety of label types, including wire and cable markers, panel labels, safety and maintenance labels, laboratory labels. Brady™ BMP51/BMP53/BMP41 Label Maker Cartridge, Reflective Tape (B-584) is available in different widths and color.

  • Abrasion and chemical resistant
  • Colored write-on area

Specifications

Product Type BMP51/BMP53 Label Maker Cartridge
Color White
Material Reflective Tape
Length (English) 25 ft.
Width (English) 1.5 in.
Thickness (English) 0.0055 in.
Adhesive Type Acrylic
Format Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.