Learn More
Enzo Life Sciences BMPO (high purity) (50mg)
SDP
Supplier:  Enzo Life Sciences ALX430141M050
Encompass_Preferred
Nitrone spin trap for the specific in vivo or in vitro detection of short-lived superoxide, hydroxyl and thiyl radicals. Forms distinguishable adducts which can be measured by EPR spectroscopy. Unlike with DMPO, the superoxide adduct does not decay into a hydroxyl adduct and it has a much longer half-life (t1/2=23min). Low paramagnetic impurities. No further purification required. Alternative name: 5-tert-Butoxycarbonyl-5-methyl-1-pyrroline-N-oxide. Purity: ≥99% (HPLC). Appearance: White to pinkish crystalline solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.