Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ BMPR1A (Human) Recombinant Protein

Catalog No. 89333872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-872 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-872 Supplier Abnova™ Supplier No. H00000657G01
Add to Cart
Add to Cart

Human BMPR1A full-length ORF (NP_004320.2) recombinant protein without tag.

The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. [provided by RefSeq]

SEQUENCE: MPQLYIYIRLLGAYLFIISRVQGQNLDSMLHGTGMKSDSDQKKSENGVTLAPEDTLPFLKCYCSGHCPDDAINNTCITNGHCFAIIEEDDQGETTLASGCMKYEGSDFQCKDSPKAQLRRTIECCRTNLCNQYLQPTLPPVVIGPFFDGSIRWLVLLISMAVCIIAMIIFSSCFCYKHYCKSISSRRRYNRDLEQDEAFIPVGESLKDLIDQSQSSGSGSGLPLLVQRTIAKQIQMVRQVGKGRYGEVWMGKWRGEKVAVKVFFTTEEASWFRETEIYQTVLMRHENILGFIAADIKGTGSWTQLYLITDYHENGSLYDFLKCATLDTRALLKLAYSAACGLCHLHTEIYGTQGKPAIAHRDLKSKNILIKKNGSCCIADLGLAVKFNSDTNEVDVPLNTRVGTKRYMAPEVLDESLNKNHFQPYIMADIYSFGLIIWEMARRCITGGIVEEYQLPYYNMVPSDPSYEDMREVVCVKRLRPIVSNRWNSDECLRAVLKLMSECWAHNPASRLTALRIKKTLAKMVESQDVKI

Specifications

Accession Number NP_004320.2
Formulation 25 mM Tris-HCl of pH8.0 containing 2% glycerol.
Gene ID (Entrez) 657
Molecular Weight (g/mol) 60.2
Preparation Method in vitro wheat germ expression system with proprietary liposome technology
Purification Method None
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias 10q23del, ACVRLK3, ALK3, CD292
Species Wheat Germ (in vitro)
Protein Tag None
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.