Learn More
AdipoGen BMS-3 (25 mg)
SDP
Supplier:  AdipoGen AGCR12901M025
Encompass_Preferred
Chemical. CAS: 1338247-30-5. Formula: C17H12Cl2F2N4OS. MW: 429.3. Potent LIMK (Lim kinase) inhibitor. Has IC50 values of 5nM and 6nM for LIMK1 and LIMK2, respectively.
Catalog No. 50-205-3868