Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BMS 345541, IKK-2 and IKK-1 Inhibitor, 10 mg

Catalog No. 502583271
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3271 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3271 Supplier Abcam Supplier No. AB14482210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly selective, orally active cell-permeable IKK-2 and IKK-1 inhibitor that blocks NF-κB-dependent transcription.

  • Molecular Weight: 255.32 Da
  • Purity: >98%
  • Inhibitor Type: Highly selective cell permeable IKK-2 and IKK-1 inhibitor
  • IC50 Values: 0.3 μM for IKK-2 and 4 μM for IKK-1
  • Biological Activity: Blocks NF-κB-dependent transcription; shows anti-inflammatory and radiosensitizing effects in vivo; orally active

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 99.2% BMS 345541
Shipping Condition Ambient
Content And Storage Store at 4°C.
Target Molecule IKK-2, IKK-1
Formulation 99.2% BMS 345541
Quantity 10 mg
For Use With (Application) Inhibition of IKK-1 and IKK-2, NF-κB-dependent transcription
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.