Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BMS 345541, IKK-2 and IKK-1 Inhibitor, 50 mg

Catalog No. 502583270
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3270 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3270 Supplier Abcam Supplier No. AB14482250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly selective cell-permeable IKK-2 and IKK-1 inhibitor that blocks NF-κB-dependent transcription.

  • Molecular Weight: 255.32 Da
  • Purity: >98%
  • Inhibitor Activity: Highly selective cell permeable IKK-2 and IKK-1 inhibitor (IC50 values are 0.3 μM for IKK-2 and 4 μM for IKK-1)
  • Biological Effect: Blocks NF-κB-dependent transcription
  • In Vivo Effects: Shows anti-inflammatory and radiosensitizing effects
  • Route of Administration: Orally active
  • CAS Number: 445430-58-0

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule IKK-2, IKK-1
Shipping Condition Ambient
Content And Storage Store at 4°C.
Form Solid
Quantity 50 mg
For Use With (Application) IKK-2 inhibition, IKK-1 inhibition, NF-κB-dependent transcription inhibition, anti-inflammatory, radiosensitizing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.