Learn More
STEMCELL Technologies BMS 493, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73974
Encompass_Preferred
BMS 493 is an inverse agonist of the pan-retinoic acid receptors (RARs). RARs are ligand-dependent transcription factors that regulate gene networks that control cell growth, differentiation, survival, and death (Germain et al.). BMS 493 has been shown to increase nuclear receptor co-repressor (NCoR) interaction with RARs (Germain et al.; Rinkevich et al.).DIFFERENTIATION· Affects development of bronchial tubule formation in mice (Chazaud et al.).· Inhibits activation of immature dendritic cells (Geissmann et al.).CANCER RESEARCH · Inhibits differentiation of leukemic blasts (Kamashev et al.).
Catalog No. 50-197-6158
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.