Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 566419
GREENER_CHOICESDP

Catalog No. 549250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
549250 50 mg
549210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 549250 Supplier Tocris Bioscience™ Supplier No. 5492/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inosine monophosphatase dehydrogenase (IMPDH) inhibitor

BMS 566419 is a potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.

Specifications

Chemical Name or Material N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide
CAS 566161-24-8
Quantity 50 mg
Target Dehydrogenase Inhibitors
Molecular Formula C28H30FN5O2
Formula Weight 487.57
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.