Learn More
Apexbio Technology LLC BMS-707035 729607-74-3 100mg
SDP
Supplier:  Apexbio Technology LLC A4069100
Encompass_Preferred
BMS-707035 (CAS 729607-74-3) is a small-molecule inhibitor targeting HIV-1 integrase It is designed to block integrase-mediated DNA strand transfer thereby inhibiting the integration of viral DNA into host genomic DNA BMS-707035 exerts its biological activity primarily through specific binding to HIV-1 integrase interfering with the interaction between integrase and target DNA In in vitro studies BMS-707035 demonstrates inhibitory activity with a reported IC50 value of approximately 15 nM Based on these pharmacological properties BMS-707035 holds research potential in studies of HIV-1 integration mechanisms integrase inhibitor resistance and integrase-DNA interaction dynamics
Catalog No. 50-000-93203
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.