Learn More
Selleck Chemical LLC BMS303141 S0277-5MG
SDP
Supplier:  Selleck Chemical LLC S02775MG
Encompass_Preferred
BMS-303141 is a potent cell-permeable inhibitor of ATP-citrate lyase (ACL) with IC50 of 0 13 M BMS-303141 shows inhibition of total lipid syntheses with IC50 of 8 M in HepG2 cells
Catalog No. 50-217-3030
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.