Learn More
Genscript Corporation BNP 14D5 mAb Mouse
Supplier:  Genscript Corporation V00602100
Encompass_Preferred
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction It can be used for diagnosis of congestive heart failure (CHF)
Catalog No. 50-000-56746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.