Learn More
Bachem BNP-32 (human) Acetate
SDP
Supplier:  Bachem 4016567.1000
Encompass_Preferred
BNP-32 (human) Acetate, H-Ser-Pro-Lys-Met-Val-Gln-Gly-Ser-Gly-Cys-Phe-Gly-Arg-Lys-Met-Asp-Arg-Ile-Ser-Ser-Ser-Ser-Gly-Leu-Gly-Cys-Lys-Val-Leu-Arg-Arg-His-OH, 124584-08-3, C143H244N50O42S4, Mr 3464.09, 1mg. Nesiritide, Brain Natriuretic Peptide-32 (human), BNP (1-32), human. The secretion of the cardiac hormone BNP-32 is increased considerably in patients with congestive heart failure, successful medical treatment of this condition will result in a decrease. Yandle et al. described a method for determining hBNP in human plasma. Effects of hBNP infusion in humans have been described by Holmes et al and La Villa et al. BNP is an important biomarker in the diagnosis of heart diseases. Additionally, BNP-32 may act as a neuropeptide. BNP-32 exerts strong lipolytic effects in humans.
Catalog No. 50-260-219
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.