Learn More
Description
Specifications
Specifications
| Antigen | BNP |
| Applications | ELISA, Immunoassay, Sandwich ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 50E1cc |
| Conjugate | DyLight 650 |
| Dilution | Western Blot, ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein |
| Host Species | Mouse |
| Immunogen | Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
