Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598295
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598295 Supplier Invitrogen™ Supplier No. PA598295
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BNP Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is a member of the natriuretic peptide family and encodes a secreted protein which functions as a cardiac hormone. The protein undergoes two cleavage events, one within the cell and a second after secretion into the blood. The protein's biological actions include natriuresis, diuresis, vasorelaxation, inhibition of renin and aldosterone secretion, and a key role in cardiovascular homeostasis. A high concentration of this protein in the bloodstream is indicative of heart failure. Mutations in this gene have been associated with postmenopausal osteoporosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Nppb
Gene Accession No. P16860
Gene Alias 5 kDa cardiac natriuretic peptide; AA408272; Bnf; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP-32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; type-B natriuretic factor
Gene Symbols Nppb
Host Species Rabbit
Immunogen Recombinant Human Natriuretic peptides B protein (27-102AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4879
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.