Learn More
Cayman Chemical Boc-Orn Fmoc-OH 1g
SDP
Supplier:  Cayman Chemical 304951 G
Encompass_Preferred
A steroid glycoside with diverse biological activities inhibits the replication of MHV-68 (IC50 2 77 M) and the lytic replication of human gammaherpes virus in BC3 cells reduces levels of NO PGE2 ROS and pro-MMP-9 in LPS-stimulated RAW 264 7 cells reduces ROS production and MMP-9 and MMP-2 activity in HaCat human keratinocytes induces tumor cell apoptosis and reduces tumor growth in an H22 hepatocellular carcinoma mouse xenograft model at 4 mg/kg
Catalog No. 50-299-1656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.