Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-Tyr 2-Br-Z -OH, Advanced ChemTech

Catalog No. 01340860
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-860 5 g
01-340-861 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-340-860 Supplier Advanced Chem Tech Supplier No. BY25625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 47689-67-8
Molecular Formula C22H24BrNO7
Molecular Weight (g/mol) 494.34
InChI Key UYWMYJQSTUVRHR-GOSISDBHSA-N
IUPAC Name (2R)-3-[4-({[(2-bromophenyl)methoxy]carbonyl}oxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(OC(=O)OCC2=CC=CC=C2Br)C=C1)C(O)=O

Specifications

Color White/Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Boc-Tyr 2-Br-Z -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.