Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Boc-Val-Leu-Gly-Arg-pNA Acetate Salt

Catalog No. MP08613082 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08613082 100 mg
MP08613081 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP08613082 Supplier MP Biomedicals™ Supplier No. 08613082

Boc-Val-Leu-Gly-Arg-pNA is a chromogenic substrate for horseshoe crab clotting enzyme for endotoxins detection.

  • Boc-VLGR-pNA acetate salt (sequence Boc-Val-Leu-Gly-Arg-pNA), is a chromogenic substrate for horseshoe crab clotting enzyme
  • It has been widely used in detection assays for bacterial endotoxins
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Format Powder
For Use With (Application) Endotoxin detection
Molecular Weight (g/mol) 663.78
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.