Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY FL Polyclonal Antibody
GREENER_CHOICE

Catalog No. A5770
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5770 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5770 Supplier Invitrogen™ Supplier No. A5770
  • $552.00
    $414.65 / Each of 1

    Save $137.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. These anti-dye antibodies, including our antibody to the BODIPYTM FL fluorophore, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes. The anti-BODIPYTM FL antibody (A-5770) was raised against the BODIPY FL fluorophore and exhibits no appreciable cross-reaction with non-BODIPY dyes, although it will cross-react to varying extents with other BODIPY dyes. In solution assays, we have found that this antibody effectively quenches the fluorescence of the BODIPY FL fluorophore, but quenches the BODIPY TR dye to a lesser extent and does not significantly quench the BODIPYTM TMR dye. Invitrogen uses a sensitive quenching assay to ensure that this antibody is provided at a consistently high titer value. As supplied, 20 μL of the antibody solution is certified to produce ≥50% of the maximal fluorescence quenching of 1 mL of a 50 nM solution of BODIPY FL dye, assayed in 100 mM sodium phosphate, pH 8.0. Maximal quenching for BODIPY FL dye is ~80% of the fluorescence of the free dye. Due to steric hindrance, maximal fluorescence quenching of the BODIPY FL fluorophore covalently bound to protein may be significantly less. It is a good practice to centrifuge protein conjugate solutions briefly in a mi...

BODIPY™ FL dye is a bright, green-fluorescent dye with a high extinction coefficient and fluorescence quantum yield. In contrast to the highly water soluble fluorophores Alexa Fluor™ 488 dye and fluorescein (FITC), BODIPY FL dyes have unique hydrophobic properties ideal for staining lipids, membranes, and other lipophilic compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BODIPY FL
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias BodipyFL
Host Species Rabbit
Immunogen Raised against the BODIPY™FL fluorophore and exhibits no appreciable cross-reaction with non-BODIPY™dyes, although it will cross react to varying extents with other BODIPY™dyes.
Purification Method IgG fraction
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.