Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Body Filter 95+ Coveralls with Hood and Boots

Catalog No. 17184159 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-159 X-Large
17-184-160 Medium
17-184-161 Large
17-184-164 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-184-159 Supplier International Enviroguard™ Supplier No. 4014XL
Add to Cart
Add to Cart

Disposable coveralls are designed for environments where high levels of particulate and dust protection are needed. Proprietary, breathable fabric filters down to 0.3 microns at up to 99% efficiency.

  • Durable coveralls resist abrasions and keep you cool over long hours
  • Rest assured, Body Filter 95+ Coveralls are designed to withstand the toughest working environments
  • Attached hood and boots protect the head, neck, and lower legs for full-body protection
  • Filters out 95 to 99% of 0.3 micron particles, similar to an N95 respirator
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas
  • A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
  • Elastic wrists help keep sleeves in place to prevent skin exposure
  • Possible applications include mold remediation, manufacturing, industrial applications, auto body painting, spray painting applications, and more

Specifications

Product Type Single-use Coveralls
Size X-Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Abrasion, Particle
Latex Free Yes
Model 4014
No. of Pockets None
For Use With (Application) Pharmaceuticals, Healthcare, Manufacturing, Industrial, Cleanrooms, Automotive, Overspray, Mold Remediation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.