Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boiling Stones, Solstice

Catalog No. 60047690 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-690 Supplier Solstice Supplier No. 85106500G
Add to Cart
Add to Cart

sieve analysis > 1 mm, >95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boiling Stones
Quantity 500 g
Melting Point 2038°C
pH 9.4 to 10.1 (at 20°C)
Boiling Point 2980°C
CAS 1344-28-1
Packaging Poly Bottle
UN Number NONH for all modes of transport
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.