Learn More
Invitrogen™ Bolt™ Bis-Tris Welcome Pack, 12%
Description
The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes.
The Welcome Pack contains:
- Mini Gel Tank (A25977)
- Bolt Bis-Tris Plus gels (2 boxes, 20 gels)
- Bolt MES Running Buffer, 20X (B0002)
- MES and MOPS Running Buffer, Powder (NP000105, NP000205)
- Bolt LDS Sample Buffer, 4X (B0007)
- Bolt Sample Reducing Agent, 10X (B0009)
- PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes. Run times may vary depending on buffer conditions and power supplies used.
About Bolt Bis-Tris Plus Gels
Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8, 10, 12, and 4-12% gradient and multiple well formats. Bolt Bis-Tris gels feature wedge-shaped wells, which can easily load up to two times more sample volume allowing for detection of proteins in very dilute samples or visualization of low-abundance proteins.
For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Warranty and Services
Up to 12 months
Specifications
Specifications
| Gel Percentage | 12% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Bis-Tris |
| Separation Range | 3.5 to 80 kDa |
| Wells | 12-well |
| Quantity | 1 Welcome Pk. kit |
| Shipping Condition | Approved for shipment at room temperature and wet ice |
| Product Line | Bolt, WedgeWell |
| For Use With (Equipment) | Mini Gel Tank |
Frequently Asked Questions (FAQs)
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.
- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.
- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.