Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bolt™ Empty Mini Gel Cassette Combs, 10-well

Catalog No. NW3010
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 combs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NW3010 20 combs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NW3010 Supplier Invitrogen™ Supplier No. NW3010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used with Bolt™ Empty Minigel Cassettes (Mfr. No. NW2010) for use in gel electrophoresis. Bolt™ Minigel Cassette Combs give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank.

Bolt Empty Gel Cassette Combs are for use with Bolt Empty Gel Cassettes, which give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. All cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well design. The cassettes and combs are available in the standard 1-mm thickness. The combs are available for 10 or 12 wells.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mini Gel Tank and XCell SureLock Mini-Cell
Gel Size Mini (8 x 8 cm)
Product Line Bolt
Type Empty Mini Gel Cassette Comb
Quantity 20 combs
Shipping Condition Room Temperature
Do you offer empty cassettes and combs for casting Mini and Midi protein gels?

We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).

- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.