Learn More
IMEB, Inc Pathology Bone Band Saw with Standard Blade, Attached Mounting Board, Straight Edge Guide and Foot Switch, Small Table-Top Unit
SDP
Catalog No. NC0376687
Encompass
The IMEB, Inc. Bone Band Saw is an efficient & economical way of cutting bone in the laboratory. It is only 19" high & has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust & particles. 1 Year Warranty. Lightweight & easy to clean. Portable table-top unit has a small footprint, just 17" x 13". Water-cooled diamond blade reduces dust & particles. Saw will cut bone specimens up to 3" in height; blade guide is easily adjusted. Strong Blade that can cut ultra-thin bone slice. The saw is made of aluminum & stainless steel for durability that counts! 4 blade types & added accessories are available to provide you with a versatile lab bone cutting station. *Not OSHA compliant and not NRTL tested*
Catalog No. NC0376687 Supplier IMEB, Inc Supplier No. BBS82203
The IMEB, Inc. Bone Band Saw is an efficient & economical way of cutting bone in the laboratory. It is only 19" high & has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust & particles. 1 Year Warranty. Lightweight & easy to clean. Portable table-top unit has a small footprint, just 17" x 13". Water-cooled diamond blade reduces dust & particles. Saw will cut bone specimens up to 3" in height; blade guide is easily adjusted. Strong Blade that can cut ultra-thin bone slice. The saw is made of aluminum & stainless steel for durability that counts! 4 blade types & added accessories are available to provide you with a versatile lab bone cutting station. *Not OSHA compliant and not NRTL tested*