Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bone marrow stromal cell antigen 1/CD157 Antibody Pair [HRP]
SDP

Catalog No. NB010165 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010165 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010165 Supplier Novus Biologicals™ Supplier No. NBP27956315PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse Bone marrow stromal cell antigen 1/CD157.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Gene ID (Entrez) 683
Gene Symbol BST1
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL rabbit anti-mouse Bone marrow stromal cell antigen 1/CD157 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse Bone marrow stromal cell antigen 1/CD157 monoclonal antibody,
  • Standard: Each vial contains 65 ng of recombinant mouse Bone marrow stromal cell antigen 1/CD157.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Bone marrow stromal cell antigen 1/CD157
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.